Would you like to log in again?
Break-over Wrenches with Fixed Heads, Mountz
Supplier: Mountz
06002C-110 06004D-110 06002C-109
76253-594EA 785.48 USD InStock
76253-594 76253-592 76253-590
Break-over Wrenches with Fixed Heads, Mountz
Wrenches
These wrenches are designed and manufactured to meet or exceed the accuracy and repeatability requirements of ISO 6789:2003 (6% of setting).
  • Small and lightweight
  • Tamper-proof internal adjustment
Breaks-over when preset torque is reached eliminating overtorquing. The break-over mechanism minimizes shock to assemblies. No external adjustment scale must be preset using a torque analyzer. When the TB wrench achieves torque, the head breaks-over preventing the operator from overtorquing the fastener.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty