Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stereo Zoom Trinocular Microscope, Aven Inc.

Supplier: Aven
26800B-323
76253-650EA 11274.8 USD InStock
76253-650
Stereo Zoom Trinocular Microscope, Aven Inc.
Microscopes
DSZV-44 stereo zoom trinocular microscope mounted on stand PLED featuring 60 energy saving eco friendly LED lights with dimmer.

  • Magnification range: 10x-44x
  • Zoom ratio: 4.4:1
  • Field of view: 23mm-5.2mm (0.91”-0.2”)
  • Working distance: 90mm (3.54”)
  • Includes wide field DSW10x (F.N.23) eyepieces
  • Diopter adjustment on both eyepiece tubes
  • Adjustable interpupillary distance of 52mm-75mm (2.04”-2.95”) accommodates any operator

A dual view trinocular port allows viewing of the object through the eyepieces and external monitor at the same time.

Mighty Cam USB 5M CMOS Camera With ezMeasure Software allows view to easily capture images onto to your PC for documentation, sharing or measurement.

Superior optics with crystal clear high resolution images.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty