Would you like to log in again?
Conductive Diamond™ Anti-Fatigue Industrial Floor Mats, Ranco Industries
Supplier: Ranco Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Conductive Diamond™
ECD-36X18 ECD-2436 SBD-36X2
76253-700EA 232.07 USD InStock
76253-700 76249-710 76252-072
Conductive Diamond™ Anti-Fatigue Industrial Floor Mats, Ranco Industries
Floor Mats Traction and Anti-Fatigue Floor Mats
Conductive anti-fatigue industrial floor mat disperses static electricity from an operator the moment they step on the mat and safely away from sensitive equipment.
  • Electrically conductive diamond plate mat surface of 5/32" thick with a resilient closed cell polymeric sponge base.
  • With beveled edges
  • Resistivity >106 OHMS, test method ESD-S7.1 1994
  • Conductive anti-fatigue matting should always be used with a ground cord
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty