Would you like to log in again?
Circular Connectors, PT Series, Newark/Element
Supplier: Newark
PT06A10-6P(SR) PT06A-10-6P
76253-732EA 110.93 USD InStock
76253-732 76253-730
Circular Connectors, PT Series, Newark/Element
Electrical Sockets
The PT series miniature cylindrical connectors offer high density contact arrangements in a circular shell with salt spray exposure of 48 hour, 500 hour. These connectors have 5 key/keyway polarization with 3 point bayonet lock coupling for positive mating with no chance of cross plugging. Spring tension provided by a wave washer in the coupling nut ensures maintenance of interfacial seal between mating halves. Both the insert and main joint gasket are moulded from resilient neoprene insulator.
  • Quick disconnect bayonet coupling for rapid mating and unmating
  • Operating temperature range from –55 to 125 °C
  • Copper alloy solder pin contacts with gold plating
  • Aluminium alloy body material with olive drab chromate over cadmium plating
These connectors are designed for harsh environments and have well-proven electrical capability to withstand extreme shock, exposure and vibration. The high reliability performance makes PT series connectors ideal for environmental sealing or pressurized applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty