Would you like to log in again?
Tentube™ Couplings, Colder Products Company
Supplier: CPC (Colder Products Company)
TFB1002 TMB1002
76267-602BX 314.4 USD InStock
76267-602 76253-786
Tentube™ Couplings, Colder Products Company
Fittings and Connectors
Tentube™ coupling provides one easy-to-use quick disconnect for up to ten separate fluid lines.
  • Valves on the body side in panel mount or in-line configurations for maximum design flexibility
  • Connect and disconnect up to ten lines with the use of a simple slide latch
Tubing orientation is ensured by keying the coupling body and insert. Different fluids and tubing sizes can be accommodated by using acetal or polypropylene inserts in three popular sizes.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty