Would you like to log in again?
Spot Remover, Orange-Sol Group
Supplier: Orange-Sol
40184
76253-910CS 804.93 USD InStock
76253-910
Spot Remover, Orange-Sol Group
Cleaning Agents Detergents and Surface Cleansers
Organic, biodegradable, environmentally friendly, non-toxic cleaner for low residue cleaning and degreasing on electronics and electrical cleaning.
Excellent water-dilutable, non-flammable replacement for chlorinated solvents and other harsh ink removers. In addition this clear formula will also clean asphalt, chewing gum, cosmetic stains, grease, oil, soap scum and tar.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty