Would you like to log in again?
Sanitizers, Sunburst Chemicals
Supplier: Sunburst
683505
76254-002EA 533 USD InStock
76254-002
Sanitizers, Sunburst Chemicals
Cleaning Agents Detergents and Surface Cleansers
Lower cost rinse alternative that delivers results regardless of conditions or equipment type.
  • Liquid dish machine drying agent
  • Effective in all water conditions
  • Leas glassware clear and film free
Formulated for low and high-temperature machines.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty