Would you like to log in again?
Scotch® Super 33+™ Electrical Tapes, Vinyl, 3M™
Supplier: 3M
Scotch®
7010349613 7000043003
76242-412CS 2524.89 USD InStock
76242-412 76254-076
Scotch® Super 33+™ Electrical Tapes, Vinyl, 3M™
Tapes
Scotch® Super 33+™ Vinyl Electrical Tapes are premium grade, flame-retardant all-weather vinyl-insulating tape for cable splices up to 600 V.
  • Tensile strength rating of 15 lbs. per inch meets professional electrical standards
  • Compatible with rubber and synthetic splicing compounds
  • Made of black durable PVC
These abrasion-resistant tape offers electrical and mechanical protection with the combination of an aggressive, rubber-resin adhesive and an elastic PVC backing. This tape is highly resistant to UV rays, moisture, alkalis, acids, corrosion and varying weather conditions.
These are ideal choice for providing a protective jacket for low- and high-voltage buses, as well as harness cables/wires. This tape is compatible with solid, dielectric cable insulations, rubber and synthetic splicing compounds, as well as epoxy and polyurethane resins.
Certifications: UL listed, ASTM approved, CSA certified and RoHS 2011/65/EU compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty