Would you like to log in again?
Miniature and Micro Miniature Brushes, Brass, Tanis Inc
Supplier: Tanis
04986 04994 4988 05190
76254-334PK 109.59 USD InStock
76254-334 76254-268 76254-336 76254-340
Miniature and Micro Miniature Brushes, Brass, Tanis Inc
Brushes
Miniature brushes with loop for hand use. Single stem and spiral loop end.
  • Stem Ø 0.62"
  • Filament Ø 0.003"
Used for ultra-fine cross hole deburring, cleaning, blending and polishing of small diameter holes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty