Would you like to log in again?
Labx® 123 100% Manila Hemp Nonwoven Cleanroom Wipers, Berkshire
Supplier: Berkshire
LB123.0909.60
76254-352CS 1795.22 USD InStock
76254-352
Labx® 123 100% Manila Hemp Nonwoven Cleanroom Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Labx® 123 is an environmentally friendly wipe that has been designed for ISO Class 6 (Class 1000) and above cleanroom applications.
  • Nonabrasive
  • Low extractables
  • High absorbency to weight ratio
These wipes are 100% biodegradable, and are made from 100% manila hemp. Labx® 123 is specifically engineered as a lightweight and low lint material.
Certifications: ISO Class 6 to 8. Cleanroom Class 1000 to 100000
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty