Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Vikan® Round Scoops, Remco Products
Supplier: Remco
56816 56823 56818 56814 56817 56812 56822 56824 56811 56825 56826 56815 56813
76254-426EA 21.78 USD InStock
76254-426 76254-436 76254-428 76254-438 76254-422 76254-444 76254-432 76254-424 76254-446 76254-434 76254-440 76254-442 76254-430
Vikan® Round Scoops, Remco Products
Scoops
These full color-coded bowl scoops are great for measuring and scooping liquids.
  • Ergonomically designed round bowl
  • Lightweight and durable
  • Easy to clean
  • Autoclavable at 121 °C (250 °F)
These are designed with a pouring spout on two sides to enable both left and right-hand users to pour from the scoop. The one-piece construction eliminates areas where bacteria can hide. Smooth surface makes it extremely easy to clean, and ideal for use in food production areas for moving food ingredients, liquids etc. There is a measuring scale inside the scoop.
Certifications: FDA Compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty