Would you like to log in again?
Oakton® EcoTestr™ Salinity Waterproof Pocket Tester, Cole-Parmer
Supplier: Cole Parmer
Oakton® EcoTestr™
35634-12
76258-438EA 128.52 USD InStock
76258-438
Oakton® EcoTestr™ Salinity Waterproof Pocket Tester, Cole-Parmer
Electrochemical Meters Salinity Meters
Designed to be durable and easy to use.
  • View measurements clearly on large LCD
  • Rugged housing is waterproof, dustproof, and floats in water
  • Secure, attach a lanyard to prevent losing or dropping in the field
  • Auto shutoff extends battery life
  • Two years warranty
Leakproof cap with fill line functions as a sample/calibration and hydrating cup.
Certifications: CE marked, IP 67 rated housing.
Delivery information: Includes plastic storage case, lanyard, and four AAA batteries.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty