Would you like to log in again?
Oakton® pHTestr® 5 Waterproof Pocket Testers, Cole-Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
35634-52 35634-46 35634-16
76258-468EA 437.15 USD InStock
76258-468 76258-450 76258-440
Oakton® pHTestr® 5 Waterproof Pocket Testers, Cole-Parmer
Electrochemical Meters pH and Multi-Parameter Meters
Save money by replacing sensor, not the tester.
  • Large easy-to-read LCD features three-color backlight
  • Simple to calibrate with automatic standard recognition
  • Rugged housing is waterproof, dustproof, and floats in water with sensor cap on
  • Leakproof cap with fill line functions as a sample/calibration and hydrating cup
  • Secure - attach a lanyard to prevent losing or dropping in the field
  • Auto shutoff extends battery life
  • Two years warranty
NIST and USA buffer recognition.
Certifications: CE marked, IP67 rated housing.
Delivery information: Includes plastic storage case, lanyard, and four AAA batteries.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty