Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Leptin Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
210-401-320S
76262-628EA 148 USD InStock
76262-628
Anti-Leptin Rabbit Polyclonal Antibody
Antibodies
Anti-Leptin antibody detects leptin protein. Leptin may function as part of a signaling pathway that acts to regulate the size of the body fat depot. An increase in the level of LEP may act directly or indirectly on the CNS to inhibit food intake and/or regulate energy expenditure as part of a homeostatic mechanism to maintain constancy of the adipose mass. Obesity associated leptin is the product of the OB gene and has been identified with Type II diabetes. Anti-leptin Antibody is ideal for investigators involved in Cell Signaling, Obesity research, aging research and Signal Transduction research.

This IgG fraction of anti-Mouse Leptin was tested for use in ELISA and immunoblotting.  Reactivity in other immunoassays is unknown. The antibody will recognize recombinant and native 16 kDa leptin from mouse and human.

Type: Primary
Antigen: Leptin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty