Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HER2 Recombinant Monoclonal Antibody

Supplier: Rockland Immunochemicals
400-001-GY3
76262-634EA 534 USD InStock
76262-634
Anti-HER2 Recombinant Monoclonal Antibody
Antibodies
HER2 (human epidermal growth factor receptor 2) is a member of the epidermal growth factor receptor (HER/EGFR/ERBB) family. It is also called ERBB2, CD340 or proto-oncogene Neu. The protein is a receptor tyrosine kinase located on the plasma membrane of cells. Activation of the tyrosine kinase promotes cell proliferation and suppresses apoptosis. HER2 can dimerize with any other member of the ErbB family, which results in auto phosphorylation of tyrosine residues and activation of signal pathways. HER2 is over expressed in a significant proportion of cancer patients, resulting in an increased disease recurrence and poor prognosis. Therefore an early diagnosis of the HER2 status of the tumors is necessary to decide the appropriate treatment. Besides in breast cancer HER2 is over expressed in variety of different tumors. The over expression of HER2 makes it a target for immunotherapy of certain tumor patients. The antibody Trastuzumab (Herceptin) which recognizes HER2 was approved for the treatment of HER2 positive breast cancer patients in 1998. The original approval was for treatment of HER2 positive metastatic breast tumors. Later studies have shown that Herceptin is also beneficial for early stage HER2 positive tumor patients and it is approved as an adjuvant treatment. Additionally the antibody is now approved also for other HER2 positive tumors (metastatic gastric adenocarcinoma). This antibody is made from a humanized Fab fragment making it specific, efficient, and effective. Humanized Recombinant Anti-HER2 Fab fragment Antibody is useful for researchers interested in Cancer research.

Humanized Recombinant Anti-HER2 Fab fragment Antibody has been tested for use in Flow Cytometry, Western Blot, and ELISA. This antibody recognizes structured HER2 and will work in western blot when the protein has not been denatured with DTT or bMe. Although not tested, this antibody could be useful in in IHC and in in-vivo and other cellular assays. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: HER2
Clonality: Recombinant Monoclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty