Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ccnd3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GN2S
76262-662EA 148 USD InStock
76262-662
Anti-Ccnd3 Rabbit Polyclonal Antibody
Antibodies
Anti-Cyclin D3 antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Cyclin D3 belongs to a highly conserved cyclin family, whose members are the ultimate recipients of oncogenic signals. Cyclin D3 is a key component of the cell cycle progression machinery and induces progression through the G1 phase of the cell cycle. Cyclin D3 is expressed in nearly all proliferating cells, and shows the most broad expression pattern of all three D-type (D1-D3) cyclins. Cyclin D3 is encoded from the 6p21 chromosome region and the protein is predominantly localized in the nucleus. Once induced, cyclin D3 binds and activates its associated cyclin-dependent kinases CDK4 and CDK6. Amplification of the cyclin D3 gene and overexpression of cyclin D3 protein is seen in several human cancers. A large number of human malignancies contain lesions in pathways impacting on cyclin D3. Abnormal expression of Cyclin D3 is believed to be a driving force in several human cancers. A possible role for cyclin D3 in the malignancies of the lymphoid system is suggested by the observations that cyclin D3 gene is rearranged in several neoplastic diseases such as diffuse large B cell lymphomas or multiple myelomas. Anti-Cyclin D3 is ideal for researchers interested in Cancer Research and Immunology research.

This affinity purified antibody has been tested for use in ELISA, immunohistochemistry, flow cytometry, and by immunoprecipitation. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 32.4 kDa in size corresponding to Cyclin D3 protein by western blotting in the appropriate stimulated tissue or cell lysate or extract. This reagent reacts equally to phosphorylated and non-phosphorylated Cyclin D3 by ELISA and western blotting.

Type: Primary
Antigen: Ccnd3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty