Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-F11R Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GN5
76262-668EA 518 USD InStock
76262-668
Anti-F11R Rabbit Polyclonal Antibody
Antibodies
Anti-JAM A pY280 was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). JAM A pY280 Antibody detects junctional adhesion molecule A (JAM A) protein phosphorylated at the tyrosine 280 location. JAM A is involved in cell-cell contact and tight junction formation in epithelial cells. JAM A protein is a transmembrane protein expressed in epithelial and endothelial cells as well as in hematopoietic cells and is important for a variety of cellular processes including tight junction assembly, leukocyte transmigration, platelet activation, angiogenesis and virus binding. In addition, JAM A may be a potential therapeutic target for breast cancer.

This affinity purified antibody has been tested for use in ELISA, IF, and western blot.   Specific conditions for reactivity should be optimized by the end user. Expect a band ~ 32.5 kDa in size corresponding to JAM A by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: F11R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty