Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Pax7 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GR7S
76262-704EA 148 USD InStock
76262-704
Anti-Pax7 Rabbit Polyclonal Antibody
Antibodies
The Anti-Pax7 antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Paired box protein Pax-7 transcription factor plays a role in myogenesis through regulation of muscle precursor cells proliferation. These genes play critical roles during fetal development and cancer growth. It is expressed specifically during the development of the nervous and muscular system. Anti-Pax7 antibody is ideal for researchers interested in developmental biology, stem cell research, cell growth and cancer research.

Anti-Pax7 antibody has been tested in ELISA and Western Blot.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 54.9 kDa in size corresponding to Pax7 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: Pax7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty