Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC7A11 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GU3S
76262-708EA 148 USD InStock
76262-708
Anti-SLC7A11 Rabbit Polyclonal Antibody
Antibodies
xCT belongs to the amino acid-polyamine-organocation (APC) superfamily and L-type amino acid transporter (LAT) family. xCT gene encodes a member of a heteromeric, sodium-independent, anionic amino acid transport system that is highly specific for cysteine and glutamate. In this system, designated Xc(-), the anionic form of cysteine is transported in exchange for glutamate. Increased expression of this gene results in neuronal cell death. xCT may be associated with kaposi sarcoma, dyscalculia, cystinuria, spondylolsis, and anemia of prematurity. Anti-xCT Antibody is useful for researchers interested in Cancer Research, Glucose/Energy Metabolism Research, and Cell Surface Protein Interaction Research.

Anti-xCT Antibody has been tested in Western Blot, ELISA, Immunofluorescence, and flow cytometry. Expect a band at 55.4 kDa in western blot using appropriate lysates. Positive control used HeLa, NIH-3T3, A549, HCT-116, or PC3 whole cell lysates at 0.1-1.0µg/mL for WB and PC3 with MeOH at 10µg/mL in IF. Although not tested, this antibody is likely functional in immunohistochemistry and immunoprecipitation.

Type: Primary
Antigen: SLC7A11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty