Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TP53 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GV1
76262-722EA 518 USD InStock
76262-722
Anti-TP53 Rabbit Polyclonal Antibody
Antibodies
TP53 (tumor suppressor gene p53) is one of the most well-studied genes that suppresses tumor formation and renders protection against DNA damage by inducing cell cycle arrest, DNA repair, or apoptosis. TP53 signaling is triggered through numerous cellular events ranging from DNA damage to hypoxia, stress and a plethora of other causes. Upon activation, p53 acts as zinc-containing transcriptional regulator and initiates a cascade of events that determines the cellular outcome including cell cycle arrest, apoptosis, senescence, DNA repair, development, differentiation and tissue homeostasis. Cell cycle arrest is induced by p53 via trans-activating genes such as p21 (CDK-inhibitor 1, cyclin dependent kinase) and others. Interestingly, p53 itself is capable of triggering cellular responses (survival or induced cell death) as well. Mutations or deletions in the TP53 gene are present in nearly 50% of human cancers, and primarily results in impaired tumor suppressor function. Anti-p53 (ac Lys292) antibody is ideal for researchers interested in developmental biology, cell growth and cancer research.

p53 K292 Ac Antibody is suitable for ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~43.6 kDa in size corresponding to p53 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: TP53
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty