Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIRT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GW4S
76262-744EA 148 USD InStock
76262-744
Anti-SIRT1 Rabbit Polyclonal Antibody
Antibodies
SIRT1 antibody detects human SIRT1 protein. SIRT1 is a member of the sirtuin family of protein-modifying enzymes. SIRT1 is a NAD+-dependent deacetylase that plays an important role in many cellular processes. SIRT1 protein is known to regulate epigenetic gene silencing and suppress recombination of rDNA. SIRT1 deacetylates a wide range of substrates, including p53, NF-kB, FOXO transcription factors, and PGC-1 alpha, with roles in cellular processes such as muscle differentiation, adipogenesis, protection from axonal degeneration, and life span extension. SIRT1 is downregulated in cells that have high insulin resistance and inducing its expression increases insulin sensitivity, suggesting the molecule is associated with improving insulin sensitivity. Furthermore, SIRT1 de-acetylates and affects the activity of both members of the PGC1-alpha/ERR-alpha complex, which are essential metabolic regulatory transcription factors. Anti-SIRT1 Antibody is useful for researcher involved in research areas including cancer, diabetes, aging, neurodegenerative, metabolic or cardiovascular diseases.

This affinity purified antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band ~62 or 81.7 kDa in size corresponding to SIRT1 by western blotting in the appropriate cell lysate or extract. Higher observed band ~125 kda is due to complexing proteins.

Type: Primary
Antigen: SIRT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty