Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ASAP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GY1
76262-750EA 518 USD InStock
76262-750
Anti-ASAP1 Rabbit Polyclonal Antibody
Antibodies
This antibody is designed, produced, and validated as part of a collaboration between Rockland and the National Cancer Institute (NCI). ASAP1 (ArfGAP with SH3 Domain, Ankyrin Repeat and PH Domain 1) is a Protein Coding gene. This gene encodes an ADP-ribosylation factor (ARF) GTPase-activating protein. The GTPase-activating activity is stimulated by phosphatidylinositol 4,5-biphosphate (PIP2), and is greater towards ARF1 and ARF5, and lesser for ARF6. This gene maybe involved in regulation of membrane trafficking with cell growth or actin cytoskeleton remodeling by binding to both SRC and PIP2. ASAP1 may function as a signal transduction protein involved in the differentiation of fibroblasts into adipocytes and possibly other cell types (By similarity) and plays a role in ciliogenesis. Anti-ASAP1 is useful for researchers interested in GTPase activities and EGFR1 Signaling Pathway.

Anti-ASAP1 Antibody has been tested for use in ELISA and Western Blot. Expect a band approximately ~125kDa using positive control lysates over-expressing ASAP1, or other specific lysates or tissues. Although not tested, this antibody could be useful in Immunofluorescence or Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: ASAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty