Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COL4A4 Rabbit Polyclonal Antibody (FITC)

Supplier: Rockland Immunochemicals
600-402-106
76262-772EA 539 USD InStock
76262-772
Anti-COL4A4 Rabbit Polyclonal Antibody (FITC)
Antibodies
Anti-Collagen Type IV Fluorescein Conjugated Antibody detects collagen. Type-IV collagen is a type of collagen found primarily in the basal lamina. It is the major structural element of basal membranes. Anti-Collagen Type IV antibody is ideal for investigators involved in Cell Signaling, Neuroscience and Signal Transduction research.

Anti-Collagen Type IV Fluorescein Conjugated Antibody is suitable for western blotting, IHC and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: COL4A4
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty