Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK3 Rabbit Polyclonal Antibody (FITC)

Supplier: Rockland Immunochemicals
600-402-GP0S
76262-788EA 148 USD InStock
76262-788
Anti-MAPK3 Rabbit Polyclonal Antibody (FITC)
Antibodies
ERK1 antibodies detect the ERK1 isoform. Mitogen activiated protein kinase 3, also known as MAPK3, ERK, or ERK1, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. ERK1 and ERK2 are activated by MEK1 and MEK2 in the B-raf signaling pathway resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Human ERK1 and ERK2 are 84% identical in sequence and share common functionality in cells. Anti-ERK1 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-ERK 1 (RABBIT) antibody is suitable for use in ELISA and Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 43 kDa.

Type: Primary
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty