Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COL1A1 Rabbit Polyclonal Antibody (HRP)

Supplier: Rockland Immunochemicals
600-403-103
76262-802EA 543 USD InStock
76262-802
Anti-COL1A1 Rabbit Polyclonal Antibody (HRP)
Antibodies
COLLAGEN Type I Peroxidase Conjugated Antibody is specific for Collagen Type I. Collagen Type I (Type-I collagen) is the most abundant collagen of the human body. It is present in scar tissue, the end product when tissue heals by repair. It is also found in tendons, the endomysium of myofibrils and the organic part of bone. Anti-collagen Type I antibody is suitable for Cancer research and other general research.

Anti-COLLAGEN Type I Peroxidase Conjugated Antibody is suitable for western blot, immunoprecipitatioin, Flow Cytometry, and immunohistochemistry. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: COL1A1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty