Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COL2A1 Rabbit Polyclonal Antibody (HRP)

Supplier: Rockland Immunochemicals
600-403-104
76262-804EA 539 USD InStock
76262-804
Anti-COL2A1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Anti-Collagen Type II antibody detects Type-II collagen which is the basis for articular cartilage and hyaline cartilage. It makes up 50% of all protein in cartilage and 85-90% of collagen of articular cartilage. Type II collagen does form fibrils which allow cartilage to entrap the proteoglycan aggregate as well as provide tensile strength to the tissue. Anti-Collagen Type II antibody is ideal for investigators involved in Immunology, Neuroscience and Signal Transduction research.

Anti-Collagen Type II Peroxidase Conjugated Antibody is suitable for western blotting, IP, IHC and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: COL2A1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty