Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SERPINA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-147S
76262-926EA 148 USD InStock
76262-926
Anti-SERPINA1 Rabbit Polyclonal Antibody
Antibodies
ALPHA-1-ANTI-TRYPSIN Antibody is specific for alpha-1-anti-trypsin. Alpha 1-Antitrypsin or α1-antitrypsin (A1AT) is a protease inhibitor belonging to the serpin superfamily and is generally known as serum trypsin inhibitor. Alpha-1-anti-trypsin protects tissues from enzymes of inflammatory cells. A1AT antibody is suitbale for immunology and cardiovascular researcher.

Anti-Human ALPHA-1-ANTI-TRYPSIN Antibody has been assayed against 1.0 µg of Human Alpha-1-anti-Trypsin in a standard ELISA using Peroxidase conjugated Affinity Purified Donkey anti-Rabbit IgG and TMB as a substrate for 30 minutes at room temperature.  A working dilution of 1:10,000 to 1:50,000 is suggested for this product.

Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty