Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-268S
76262-928EA 148 USD InStock
76262-928
Anti-AKT1 Rabbit Polyclonal Antibody
Antibodies
AKT is a component of the PI-3 kinase pathway and is activated by phosphorylation at  Ser 473 and Thr 308.  AKT is a cytoplasmic protein also known as AKT1, Protein Kinase B (PKB) and rac (related to A and C kinases). AKT is a key regulator of many signal transduction pathways.  AKT Exhibits tight control over cell proliferation and cell viability.  Overexpression or inappropriate activation of AKT is noted in many types of cancer.   AKT mediates many of the downstream events of PI 3-kinase (a lipid kinase activated by growth factors, cytokines and insulin). PI 3-kinase recruits AKT to the membrane, where it is activated by PDK1 phosphorylation. Once phos-phorylated, AKT dissociates from the membrane and phos-phorylates targets in the cytoplasm and the cell nucleus.  AKT has two main roles: (i) inhibition of apoptosis; (ii) promotion of proliferation.

This antibody is phospho specific for pS473 and is suitable for western blotting, immunohistochemistry (formalin-fixed paraffin-embedded sections), immunofluorescence and ELISA. By immunoblot a single band of the expected apparent molecular weight is observed.  For immunohistochemistry no pre-treatment of sample is required.

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty