Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Abi1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GT5
76262-938EA 518 USD InStock
76262-938
Anti-Abi1 Rabbit Polyclonal Antibody
Antibodies
Anti-Abi1 Antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Abi1 gene has been suggested to have a role in inhibiting cell growth and encodes an adaptor protein in the Abelson-interactor family. The protein is involved in signal transduction, actin regulation and cytoskeletal remodeling. Studies suggest that Abi1 is a member of the WAVE complex and a dysfunctional complex may allow for the progression of prostate cancer. Anti-Abi1 is ideal for researchers interested in melanoma, colon and breast cancer and other cancer antibody research.

This affinity purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band ~ 65 kDa in size corresponding to Abi1 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: Abi1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty