Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PIK3CB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GT8S
76262-948EA 148 USD InStock
76262-948
Anti-PIK3CB Rabbit Polyclonal Antibody
Antibodies
Anti-PIK3C beta Antibody is suitable for researcher in Cancer, Immunology and Nuclear Signaling research.

Anti-PIK3C beta Antibody has been tested for use in ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 122.7 kDa in size corresponding to PI3K beta by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: PIK3CB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty