Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TAF10 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GW8S
76262-964EA 148 USD InStock
76262-964
Anti-TAF10 Rabbit Polyclonal Antibody
Antibodies
TAF10 (TATA-Box Binding Protein Associated Factor 10) is the protein that coordinates activities for the initiation of transcription by RNA polymerase II required for activities of more than 70 polypeptides. TFIID binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as co-activators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the small subunits of TFIID that is associated with a subset of TFIID complexes. Studies with human and mammalian cells have shown that this subunit is required for transcriptional activation by the estrogen receptor, for progression through the cell cycle, and may also be required for certain cellular differentiation programs. Anti-TAF10 Antibody is useful for researchers interested in Chromatin research, transcription factor activity research, and DNA binding and transcription co-activator activity research.

Anti-TAF10 Antibody has been tested for use in ELISA, Dot Blot, and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 21.7 kDa in size corresponding to TAF10 protein by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: TAF10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty