Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-cas9 Rabbit Polyclonal Antibody (DL800)

Supplier: Rockland Immunochemicals
600-445-GK0
76262-984EA 487 USD InStock
76262-984
Anti-cas9 Rabbit Polyclonal Antibody (DL800)
Antibodies
The Clustered Regularly Interspaced Short Palindromic Repeats (CRISPR) and CRISPR Associated (Cas) system was discovered in bacteria as a defense against foreign DNA, even being believed to act as a sort of prokaryotic immune system. The Cas9 protein is becoming a a useful tool in the field of genomic editing for its ability to induce site-directed double strand breaks in DNA. It can cleave almost any sequence complementary to the guide RNA and has recently been used for modifying the genome of human embryos for the first time.

Cas9 DyLight™800 Conjugated Antibody has been tested for use in ELISA, Immunofluorescence, and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band ~158 kDa in size corresponding to Cas-9 protein by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: cas9
Clonality: Polyclonal
Clone:
Conjugation: DL800
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty