Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Rorc Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GR8S
76263-048EA 148 USD InStock
76263-048
Anti-Rorc Rabbit Polyclonal Antibody
Antibodies
Anti-Nuclear receptor ROR gamma pS203 was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Nuclear receptor ROR gamma pS203 antibody detects mouse receptor ROR gamma phosphorylated at the serine 203 position. RAR-related orphan receptor gamma is a member of the nuclear receptor family of transcription factors. (ROR-gamma) is a key regulator of cellular differentiation, immunity, peripheral circadian rhythm as well as lipid, steroid, xenobiotics and glucose metabolism. ROR-gamma regulates the circadian expression of clock genes such as CRY1, ARNTL/BMAL1 and NR1D1 in peripheral tissues and in a tissue-selective manner. It is also involved in the regulation of the rhythmic expression of genes involved in glucose and lipid metabolism and is a negative regulator of adipocyte differentiation. It controls adipogenesis as well as adipocyte size and modulates insulin sensitivity in obesity. Isoform 2, ROR-gamma is essential for lymphoid organogenesis, in particular lymph nodes and Peyer's patches. ROR-gamma also plays an important regulatory role in thymopoiesis, and in inhibiting apoptosis of undifferentiated T cells. Anti-nuclear receptor ROR gamma pS203 is ideal for researcher's interested in autoimmune diseases such as psoriasis and rheumatoid arthritis, circadian rhythms, and immune system disorders.

This affinity purified antibody has been tested for use in ELISA and western blot.   Specific conditions for reactivity should be optimized by the end user. Expect a band ~ 61 kDa in size corresponding to receptor ROR gamma by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: Rorc
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty