Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Pol Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GT3
76263-050EA 518 USD InStock
76263-050
Anti-Pol Rabbit Polyclonal Antibody
Antibodies
Anti-Orf2 Antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). The multi-functional nuclear protein ORF2p is involved in reverse transcriptase and endonuclease processes. ORF2 elements are epigenetically silenced throughout embyronic development and it is suggested that mutations of L1 would result in detrimental de novo insertions into the genome, negatively affecting gene expression.  L1 elements are critical components in the mammalian genome, and much is still not known regarding these proteins. Studies implicate that line-1 is activated in fetal oocytes and  L1ORF2p may contribute to the eradication or selection of the oocytes.

This affinity purified antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band ~150 kDa in size corresponding to Orf2 by western blotting in the liver cell lysates or extract.

Type: Primary
Antigen: Pol
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty