Would you like to log in again?
UniStore™ Reagent Bottles, Narrow Mouth, PP
Supplier: United Scientific Supplies
UniStore™
33301 33303-BULK 33254-BULK 33302-BULK 33304-BULK 33303 33304 33301-BULK 33305 33254 33302 33253 33253-BULK 33305-BULK
76263-134PK 21.37 USD InStock
76263-134 76263-136 76263-140 76263-130 76263-142 76263-132 470339-926 470340-074 470340-042 76263-138 470338-966 470338-912 470338-968 470339-084
UniStore™ Reagent Bottles, Narrow Mouth, PP
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Translucent
  • Rigid
  • Autoclavable
UniStore™ narrow mouth PP reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty