Would you like to log in again?
Measuring Cylinder with Round Base, United Scientific Supplies
Supplier: United Scientific Supplies
CP0050 CP0010 CP1000-VWR CP0025 CP0100 CP0500 CP0250 CP2000
76263-166PK 58.64 USD InStock
76263-166 76263-178 76263-168 76263-174 76263-176 76263-180 76263-170 76263-172
Measuring Cylinder with Round Base, United Scientific Supplies
Cylinders Measuring Cylinders
Unbreakable polypropylene construction.
  • Polypropylene
  • Autoclavable
Polypropylene one-piece measuring cylinders have raised graduations for easy readability. They are autoclavable and have excellent contact clarity. The broad round base provides good stability. Stated tolerances are the same as graduated intervals below.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty