Would you like to log in again?
Separatory Funnel Holder, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
45104
76263-260PK 84.07 USD InStock
76263-260
Separatory Funnel Holder, United Scientific Supplies
Support Stands
Open design allows an unobstructed view.
  • Tapered wedges insure a firm grip
  • Front opening for an unobstructed view
This non-corrosive separatory funnel holder, molded in polypropylene, has a front opening which provides clear view of the solution and allows unobstructed placement of separating funnels. Two tapered wedges are provided to ensure firm grip on rods measuring 1/2 or 3/8" diameter.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty