Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Laboratory Scoops, United Scientific Supplies
Supplier: United Scientific Supplies
81259 81256 81258 81254 81260 81253 81257 81255
76263-298PK 45.88 USD InStock
76263-298 76263-288 76263-300 76263-294 76263-296 76263-290 76263-292 76263-062
Laboratory Scoops, United Scientific Supplies
Scoops
Conveniently designed handled scoops.
  • Autoclavable
These sturdy polypropylene scoops are available in a variety of sizes, as well as in a complete set of seven scoops. Capacities are molded into the top of each scoop for easy reference. The polypropylene material is autoclavable and will not rust or corrode.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty