Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Laboratory Trays, United Scientific Supplies
Supplier: United Scientific Supplies
81701-VWR 81702 81702
76263-312KT 35.95 USD InStock
76263-312 470338-980 76263-314
Laboratory Trays, United Scientific Supplies
Trays/Dishes/Pots Laboratory Trays/Dishes
General use tray for all laboratory needs.
  • Polypropylene
  • Autoclavable
Multipurpose, polypropylene molded trays can be used for sterilizing, drying glassware, porcelain ware, and other laboratory supplies. These steam autoclavable trays have tapered walls designed for nesting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty