Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Multi-Position Stirrers, G-Biosciences
Supplier: G-Biosciences
BT1013 BT1015 BT1014
76263-484EA 1369.07 USD InStock
76263-484 76264-008 76263-998
Multi-Position Stirrers, G-Biosciences
Stirrers Magnetic Hotplate Stirrers, Multi-position
Multi-position magnetic stirrer is designed for high-efficiency experiments.
  • Digital display of stirring speed, stir bar provides uniform agitation
  • Efficient multi-position magnetic stirrer configure 4/8/12 stirring positions
  • Stainless steel is coated with silicone resin film disk
  • It is non-slip and corrosion resistant
It efficiently uses laboratory space. DC brushless motor makes the model stable at low speed and powerful with low noise at high speed.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty