Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BioFit MVMT™ Tech Classic Cleanroom Swivel Chairs, ISO 7

Supplier: BioFit

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

MVMT™ Tech Classic
MTCL-MH-7 MTCL1FTMR7B540 MTCLTLP287P540 MTCL-MM-7 MTCL-TH-7 MTCL-TL-P28-7
76263-532EA 2211.96 USD InStock
76263-532 76263-526 76411-338 76404-374 76263-534 76263-536
BioFit MVMT™ Tech Classic Cleanroom Swivel Chairs, ISO 7
Chairs
MVMT™ stands for Movement, for ergonomic seating. The chairs are developed to address the unique ergonomic needs and working postures of users in technical environments specifically for healthcare, laboratory, cleanroom and standing-desk applications.

  • Comfortable, full ergonomic support and functionality
  • Easy, efficient and effective to clean
  • With task control to regulate seat height and backrest angle
  • Seat mounted forward for 18" seat depth
  • Lumbar support and 3" adjustable backrest height
  • 5 legged 28.7" dia solid cast aluminum base with casters, models medium and high with footring
  • Choice of mid or tall backrests

Built to withstand 3-shift use, 365 days per year and backed by a 13-year warranty (lifetime on pneumatic seat height adjustment cylinder). Proprietary QR Code Profile System provides users easy access to operational videos, warranty balance and more. Available on GSA contract.

Certifications: Cleanroom ISO 7 rated; LEED compliant; meets industry-standard ANSI/BIFMA X5.1 performance and safety guidelines.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty