Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DURAN® GL 45 Connection System, DWK Life Sciences

Supplier: DWK Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

DURAN®
1129816 1129819 1129750 1129814 1129817 1129818 1129815 1137799 1129751 1156292
76263-750PK 42.19 USD InStock
76263-750 76263-740 76263-752 76263-774 76263-768 76263-758 76263-748 76263-742 76263-754 76263-756
DURAN® GL 45 Connection System, DWK Life Sciences
Closures Bottle Closures
This GL 45 connection system is useful for safe transfer of liquid media within a closed and sterile system.

  • Flexible modular system
  • Autoclavable at 121 °C
  • Maximum temperature is 140 °C
  • Four different tubing diameters (1.6, 3.0, 3.2 and 6.0 mm) can be connected

Sterile pressure equalization is possible through use of syringe filter. Unused ports can be sealed off with a red GL 14 screw cap.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty