Would you like to log in again?
DURAN® HPLC Bottles, GL 45, DWK LIFESCIENCES
Supplier: DWK Life Sciences
DURAN®
1129820 1129821
76263-760CS 325.44 USD InStock
76263-760 76263-762
DURAN® HPLC Bottles, GL 45, DWK LIFESCIENCES
Bottles HPLC Bottles
Complete system comprising of DURAN® pressure plus laboratory bottle with four port screw cap (PP), four screw connections (black, M8 thread) and silicone seals.
  • Easy-to-read scale and large labelling field for easy marking
  • Unused ports can be sealed with silicone blanking seals
  • Autoclavable at 121 °C
Connection of different hose diameters (1.6 and 3.2 mm) as well as sterile pressure equalization sets (syringe filter 0.2 µm) is possible. It is safe to transfer of liquid media within a closed and sterile system (evaporation is reduced).
Certifications: USP standard.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty