Would you like to log in again?
DURAN® GLS 80® Quick Release Screw Caps and Pouring Rings, DWK Life Sciences
Supplier: DWK Life Sciences
DURAN®
1160166 1112716
76263-736PK 83.28 USD InStock
76263-736 76263-776
DURAN® GLS 80® Quick Release Screw Caps and Pouring Rings, DWK Life Sciences
Closures Bottle Closures
GLS 80® quick release screw caps with lip seal permits opening and closing of the bottle with only a three-quarter turn.
  • A matching pouring ring permits clean, drip-free use
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty