Would you like to log in again?
DURAN® YOUTILITY® GL 45 Laboratory Bottle System, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
218815457 218813653 218864457 218863658 218812854 218814452 218862859 218865453
76264-196CS 547.78 USD InStock
76264-196 76264-186 76264-194 76264-184 76264-188 76264-192 76264-182 76264-190
DURAN® YOUTILITY® GL 45 Laboratory Bottle System, DWK Life Sciences
Bottles Glass Bottles
The new generation of glass laboratory bottles.
  • Borosilicate 3.3 pharmacopeia type 1 neutral glass bottles
  • Bottle tags are made of silicone and available in eight colors
  • Labels accept copier, printer, ballpoint, and technical markers and inks
  • Pour rings are autoclavable hydrophilic polypropylene
The updated design includes an improved ergonomic bottle shape and dedicated accessories for better handling, sample identification, and ease of use. The GL 45 bottles have a defined labelling area and an integrated retrace code for full traceability.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty