Would you like to log in again?
DURAN® GLS 80® Double Walled Bottle, Wide Mouth, DWK Life Sciences
Supplier: DWK Life Sciences
DURAN®
242564451 242565456
76264-204CS 1997.64 USD InStock
76264-204 76264-202
DURAN® GLS 80® Double Walled Bottle, Wide Mouth, DWK Life Sciences
Bottles Glass Bottles
The DURAN® double walled, wide mouth bottles GLS 80® incorporate an integral jacket that isolates the contents from the external environment.
  • Autoclavable at 121 °C
  • Offers a sealable and more flexible alternative to open topped jacketed beakers
Heated or cooled liquids can be circulated through the jacket to control the temperature within the screw topped DURAN® bottle.
Certifications: USP standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty