Would you like to log in again?
DURAN® Aspirator (Levelling) Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
DURAN®
247035402 247036304 247037309 247038605
76264-212CS 1111.22 USD InStock
76264-212 76264-210 76264-208 76264-206
DURAN® Aspirator (Levelling) Bottles, DWK Life Sciences
Bottles Aspirators
Complete with screw connection cap, silicone seal and stopcock with PTFE spindle. Dosing of liquids is possible via a stopcock.
  • Neck with DIN thread GL 45
  • Tabulator with GL 32
  • Retrace code
  • Autoclavable
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty