Would you like to log in again?
DURAN® Gas Washing Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
DURAN®
257040101 247130008
76264-216CS 290.65 USD InStock
76264-216 76264-214
DURAN® Gas Washing Bottles, DWK Life Sciences
Bottles Gas Washing Bottles
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Autoclavable
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Wash bottle with DIN GL 45 screw pitch and adjustable trim with screw cap and two PP knobs with bolting.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty