Would you like to log in again?
DURAN® High Temperature PBT Screw Caps, DWK Life Sciences
Supplier: DWK Life Sciences
DURAN®
292400806 292401108
76264-230PK 48.15 USD InStock
76264-230 76264-232
DURAN® High Temperature PBT Screw Caps, DWK Life Sciences
Closures Bottle Closures
DURAN® red PBT closed screw caps with PTFE protected seal protect important materials in the lab.
  • High leak tightness through use of PTFE coated silicone seal
  • More chemically resistant than PP screw cap
  • Autoclavable at 121 °C
Permitting clean, drip-free use.
Ordering information: Matching ETFE pouring rings can be ordered.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty