Would you like to log in again?
Synthware Bosshead, Kemtech America
Supplier: Kemtech America
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Synthware
W200002 W200002Z J111200 W200002B
76264-414EA 19.33 USD InStock
76264-414 60005-036 76264-412 76264-416
Synthware Bosshead, Kemtech America
Clamps Bossheads
Chrome plated finish with plastic-headed screws.
  • Metal bosshead
  • Chrome plated and black metal available
  • Various tension screw designed available
Designed for use with rods ranging from 15 to 19 mm in diameter.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty